missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TARS |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16823584
|
Proteintech
14773-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16813584
|
Proteintech
14773-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Threonyl-tRNA synthetase belongs to the class-II aminoacyl-tRNA synthetase familySpecifications
| TARS | |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P26639, Q5XHY5, Q9D0R2 | |
| 110960, 294810, 6897 | |
| TARS Fusion Protein Ag6614 | |
| Primary | |
| -20°C | |
| TARS |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| TARS, Threonine tRNA ligase, threonyl tRNA synthetase, ThrRS | |
| Tars | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title