missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TARS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14773-1-AP-150UL
This item is not returnable.
View return policy
Description
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Threonyl-tRNA synthetase belongs to the class-II aminoacyl-tRNA synthetase familySpecifications
| TARS | |
| Polyclonal | |
| Unconjugated | |
| TARS | |
| TARS, Threonine tRNA ligase, threonyl tRNA synthetase, ThrRS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110960, 294810, 6897 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P26639, Q5XHY5, Q9D0R2 | |
| Tars | |
| TARS Fusion Protein Ag6614 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction