missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Taq Core Kit 10
Supplied with standard reaction buffers
£485.48 - £2553.54
Specifications
Content And Storage | Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C |
---|---|
Form | Frozen |
Format | Frozen liquid |
For Use With (Application) | Standard PCR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11417040
|
MP Biomedicals™
11EPTQK300 |
1000U |
£485.48
1 set |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11437030
|
MP Biomedicals™
11EPTQK105C |
5 x 250U |
£895.70
1250 units |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11447030
|
MP Biomedicals™
11EPTQK109 |
10 x 250U |
£1517.92
2500 units |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11457030
|
MP Biomedicals™
11EPTQK10L |
5 x 1000U |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.Specifications
Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C | |
Frozen | |
Frozen liquid | |
Standard PCR |
Research Use Only (RUO)