Learn More
MP Biomedicals™ Taq Core Kit 10
Supplied with standard reaction buffers
Brand: MP Biomedicals™ 11EPTQK10L
Code : 80
Additional Details : Weight : 0.01000kg
Includes: Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix
Description
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.
Unit definition: One unit of DNA polymerase is the amount of enzyme required to catalyse the incorporation of 10nmol of nucleosides into a DE81 adsorbable product within 30 min. at 74°C under assay conditions.Specifications
Taq DNA Polymerase (5U/μL), 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2, 10mM dNTPs mix. Store at -20°C | |
Frozen liquid | |
Standard PCR |
Frozen | |
5 x 1000U |
Research Use Only (RUO)