missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPP1/PLEKHA1 Antibody (16C9), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | TAPP1/PLEKHA1 |
|---|---|
| Clone | 16C9 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024418
|
Novus Biologicals
NBP3-26375-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032005
|
Novus Biologicals
NBP3-26375-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAPP1/PLEKHA1 Monoclonal antibody specifically detects TAPP1/PLEKHA1 in Human samples. It is validated for Western Blot, ELISASpecifications
| TAPP1/PLEKHA1 | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 59338 | |
| IgG | |
| Affinity purified |
| 16C9 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| family A (phosphoinositide bindingspecific) member 1, PH domain-containing family A member 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 1 | |
| A synthesized peptide derived from Human TAPP1/PLEKHA1 [UniProt Q9HB21] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title