missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAPP1/PLEKHA1 Antibody (16C9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26375-100ul
This item is not returnable.
Consulta la politica di reso
Descrizione
TAPP1/PLEKHA1 Monoclonal antibody specifically detects TAPP1/PLEKHA1 in Human samples. It is validated for Western Blot, ELISA
Specifica
| TAPP1/PLEKHA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 59338 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 16C9 | |
| Western Blot 1:500-1:2000, ELISA | |
| family A (phosphoinositide bindingspecific) member 1, PH domain-containing family A member 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 1 | |
| A synthesized peptide derived from Human TAPP1/PLEKHA1 [UniProt Q9HB21] | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto