missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tango™ ADRA2A-bla U2OS DA Assay Kit

Product Code. 10647574
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10647574

Brand: Thermo Scientific™ K1607

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The Tango™ ADRA2A-bla U2OS cells contain the human Adrenergic, Alpha-2A-, Receptor (ADRA2A) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

The Tango™ ADRA2A-bla U2OS cells contain the human Adrenergic, Alpha-2A-, Receptor (ADRA2A) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The Tango™ ADRA2A-bla U2OS cells cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ ADRA2A-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product. Division Arrested cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_000681.2
Assay Entry Cell-based beta-arrestin recruitment
Cell State Division-Arrested Cells
Content And Storage Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol).

Includes:
• Tango™ ADRA2A-bla U2OS DA cells
• LiveBLAzer™-FRET B/G Loading Kit, 70 μg
• Solution D
• Tango™ ADRA2A-bla U2OS DA Assay Protocol
• LiveBLAzer™-FRET B/G Loading Protocol
• Certificate of Analysis

The Tango™ ADRA2A-bla U2OS DA cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Tail from AVPR2 GPCR
Description Tango™ ADRA2A-bla U2OS DA Assay Kit, 8 x 106 Cells, ∽4,800,000 cells/vial at 4,800,000 cells/mL Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays
Gene Symbol ADRA2A
Includes Tango™ ADRA2A-bla U2OS DA assay kit Each kit contains sufficient quantities of division arrested (DA) cells and substrate to assay 1 x 384 well plate. (Other materials are required separately; please refer to the protocol). Includes: Tango™ ADRA2A-bla U2OS DA cells, LiveBLAzer™-FRET B/G loading kit, 70μg, solution D, Tango™ ADRA2A-bla U2OS DA assay protocol, LiveBLAzer™-FRET B/G loading protocol, certificate of analysis
Population Status Single Cell Clone Origin
Product Line Tango
Quantity 8 x 106 Cells
Reporter Gene BLA (Beta-Lactamase)
Sample Type U2OS
Target Entry ADRA2A
Type Division-Arrested Cells Assay Kit
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.