missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TALPID3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24421-1-AP-150UL
This item is not returnable.
View return policy
Description
KIAA0586 also named as Talpid3 is a 1472 amino-acid protein, which belongs to Talpid3 family. Talpid3 is a gene essential for vertebrate development. Studies in chicken, mouse, zebrafish and human have shown that TALPID3 is a centrosomal protein, mutants null for TALPID3 fail to produce cilia, resulting in far reaching signaling defects such as polydactyly, dorsalised neural tube, craniofacial and vascular defects often due to abnormalities in Hedgehog signal transduction. Talpid3 is absolutely required for the function of both Gli repressor and activator in the intracellular Hedgehog pathway and ciliogenesis. TALPID3 may be identified as a subcellular maker of centrosome and needed to further studied.Specifications
| TALPID3 | |
| Polyclonal | |
| Unconjugated | |
| KIAA0586 | |
| KIAA0586, KIAA0586 isoform, Talpid3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9786 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BVV6 | |
| KIAA0586 | |
| TALPID3 Fusion Protein Ag19813 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction