Learn More
Witeg™ Tall Form Glass Beakers
Product Code. 10180650
Change view
Click to view available options
Capacity (Metric):
25 mL
Unit Size:
Pack of 10
Product Code. Capacity (Metric) Unit Size
10180650 25 mL Pack of 10
1 options
Product Code. 10180650 Supplier Witeg™ Supplier No. 5 502 025 S
  • ONLINE EXCLUSIVE PRICE
    £12.75
    £11.69 / Pack of 10
    Save £1.06
    8% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
With graduations for accurate and precise filling and dispensing.
  • Tall form
  • Acc. to DIN 12331, ISO 3819
  • Manufactured from DURAN™ borosilicate glass 3.3
  • With spout for easy pouring
Specifications
Color Transparent
Capacity (Metric) 25 mL
Material Borosilicate Glass
Includes Spout
Graduated Yes
Certifications/Compliance ISO 3819, DIN 12331
Style Tall Form