Learn More
Agilent BioTek Take3 and Take3 Trio Micro-Volume Microplates
Take3 and Take3 Trio Micro-Volume plates expand functionality of the microplate spectrophotometers, multi-mode and Hybrid readers, while reducing sample volumes, costs and time required for routine quantification
Brand:  Agilent BioTek TAKE3-SN
Code : 03
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 10626785
  • £3376.34 / Each
Estimated Shipment: 01-05-2024
to see stock.
Description
Description
Measure up to 16 (Take3) or 48 (Take3 Trio) 2μL samples at a time without dilution or special treatment.
  • Compatible with Agilent BioTek Eon, Epoch, Synergy, PowerWave XS2 spectrophotometers
  • Includes Gen5 software with pre-programmed micro-volume nucleic acid and protein quantification protocols
  • Absorbance and fluorescence detection capability
  • Easy maintenance: simply wipe-off sample locations
  • Robust design
  • Take3 Trio has two locations for BioCell measurements
  • Read microspots, BioCells or standard cuvets
Take3 (11-120-571) offers:
  • 16 microspots for 2μL samples; two locations for proprietary BioCell measurements (BioCell optional) and one location for standard cuvet measurements
  • Take3 Trio (11-120-613) offers:
    • 48 microspots for 2μL samples; two locations for proprietary BioCell measurements (BioCell optional)
    Nucleic acid and protein quantification, in-situ fluorometric protein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
    Specifications
    Specifications
    Nucleic acid and protein quantification, in-situ fluorometric Pprotein quantification, 260/280 and 260/230 protein purity measurements, very low volume in-situ BCA assay, spectral scan of single or multiple low volume samples
    16
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    View accessory products