missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK1 Antibody (3E8H4), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £438.00
Specifications
| Antigen | TAK1 |
|---|---|
| Clone | 3E8H4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18210500
|
Novus Biologicals
NBP2-52509 |
0.1 mg |
£438.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675016
|
Novus Biologicals
NBP2-52509-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAK1 Monoclonal specifically detects TAK1 in Human samples. It is validated for Western Blot, ELISA.Specifications
| TAK1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| PBS and 0.5% protein stabilizer with 0.05% Sodium Azide | |
| 6885 | |
| Purified recombinant fragment of human MAP3K7 (AA: 471-579) expressed in E. Coli. | |
| Primary | |
| 67.2 kDa |
| 3E8H4 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1 | |
| MAP3K7 | |
| IgG2a | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title