missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK1 Antibody (3E8H4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52509-0.025mg
This item is not returnable.
View return policy
Description
TAK1 Monoclonal specifically detects TAK1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| TAK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1 | |
| Mouse | |
| 67.2 kDa | |
| 0.025 mg | |
| Apoptosis, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 6885 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 3E8H4 | |
| Unconjugated | |
| PBS and 0.5% protein stabilizer with 0.05% Sodium Azide | |
| MAP3K7 | |
| Purified recombinant fragment of human MAP3K7 (AA: 471-579) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction