Learn More
Cytiva Tagged Column Package, His (2-step Purification Protocol)
Combines affinity with size exclusion chromatography to obtain highly pure his-tagged proteins
Brand: Cytiva 29497630
Additional Details : CAS Number : 64-17-5 Weight : 0.04500kg
Description
- Contains a HisTrap™ HP column (1 mL) prepacked with Ni Sepharose High Performance (HP) affinity resin for preparative purification of his-tagged proteins
- Achieve final purity with HiPrep™ 16/60 Sephacryl S-200 HR size exclusion chromatography column, which removes any remaining trace of impurities or closely related substances and imidazole
- Includes instructions with guidance and tips on how to get started
Specifications
HisTrap™ HP Column Prepacked with Ni Sepharose High Performance (HP) Affinity Resin, and HiPrep™ 16/60 Sephacryl S-200 HR Size Exclusion Chromatography Column | |
1 Kit | |
Protein Purification System |
4°C to 30°C | |
Purification of His-Tagged Proteins |
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.