missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TAFA2/FAM19A2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
£147.94 - £333.68
Specifications
Antigen | TAFA2/FAM19A2 |
---|---|
Clone | 447815 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771909
|
R&D Systems
MAB4179-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18189880
|
R&D Systems
MAB4179 |
100 μg |
£333.68
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAFA2/FAM19A2 Monoclonal specifically detects TAFA2/FAM19A2 in Human samples. It is validated for Western Blot.Specifications
TAFA2/FAM19A2 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
AAH28403 | |
338811 | |
E. coli-derived recombinant human TAFA2/FAM19A2 Ala31-His131 Accession # AAH28403 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TAFA2/FAM19A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 3, 4, or 5 is observed. |
447815 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, FAM19A2, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2 | |
FAM19A2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only