missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TAFA2/FAM19A2 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB4179
Additional Details : Weight : 0.00970kg
Description
TAFA2/FAM19A2 Monoclonal specifically detects TAFA2/FAM19A2 in Human samples. It is validated for Western Blot.Specifications
TAFA2/FAM19A2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
AAH28403 | |
FAM19A2 | |
E. coli-derived recombinant human TAFA2/FAM19A2 Ala31-His131 Accession # AAH28403 | |
100 μg | |
Primary | |
Detects human TAFA2/FAM19A2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human TAFA1, 3, 4, or 5 is observed. | |
Human | |
Purified |
Western Blot | |
447815 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Chemokine-like protein TAFA-2, DKFZp761E1217, DKFZp781P0552, FAM19A2, family with sequence similarity 19 (chemokine (C-C motif)-like), member A2, MGC42403, TAFA2, TAFA-2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
338811 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only