missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TADA2B |
|---|---|
| Concentration | 0.37 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887244
|
Proteintech
17367-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877244
|
Proteintech
17367-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| TADA2B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q86TJ2 | |
| 93624 | |
| TADA2B Fusion Protein Ag10783 | |
| Primary | |
| -20°C | |
| TADA2B |
| 0.37 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ADA2 beta, ADA2 like protein beta, ADA2(beta), ADA2B, TADA2B, Transcriptional adapter 2 beta | |
| TADA2B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title