missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA2B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17367-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| TADA2B | |
| Polyclonal | |
| Unconjugated | |
| TADA2B | |
| ADA2 beta, ADA2 like protein beta, ADA2(beta), ADA2B, TADA2B, Transcriptional adapter 2 beta | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 93624 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86TJ2 | |
| TADA2B | |
| TADA2B Fusion Protein Ag10783 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction