missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACSTD2/TROP2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TACSTD2/TROP2 |
|---|---|
| Concentration | 300 μg/mL |
| Applications | Western Blot, Immunofluorescence, ELISA |
| Form | Liquid |
| Host Species | Rabbit |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18066865
|
Proteintech
29856-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020466
|
Proteintech
29856-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The TACSTD2/TROP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TACSTD2/TROP2. This antibody recognizes human antigen. The TACSTD2/TROP2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Spezifikation
| TACSTD2/TROP2 | |
| Western Blot, Immunofluorescence, ELISA | |
| Rabbit | |
| 4070 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
| 300 μg/mL | |
| Liquid | |
| Human | |
| EGP 1, GA733, GA733 1, M1S1, TACSTD2, TROP2 | |
| Primary |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts