Learn More
TACSTD2/TROP2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29856-1-AP-150UL
Description
The TACSTD2/TROP2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TACSTD2/TROP2. This antibody recognizes human antigen. The TACSTD2/TROP2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| TACSTD2/TROP2 | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| EGP 1, GA733, GA733 1, M1S1, TACSTD2, TROP2 | |
| 150 μL | |
| 4070 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |