missing translation for 'onlineSavingsMsg'
Learn More

Tacrolimus Methyl Acryl Aldehyde, TRC

Product Code. 30428433
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428433 100 mg
30420273 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30428433 Supplier Toronto Research Chemicals Supplier No. TRCT004150100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109466-74-2
Molecular Formula C11 H18 O3
Molecular Weight (g/mol) 198.26
Synonym 2-Propenal, 3-(4-hydroxy-3-methoxycyclohexyl)-2-methyl-, [1R-[1α(E),3α,4β]]- (9CI)
IUPAC Name (E)-3-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-2-methylprop-2-enal
SMILES CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)\C=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H18O3/c1-8(7-12)5-9-3-4-10(13)11(6-9)14-2/h5,7,9-11,13H,3-4,6H2,1-2H3/b8-5+/t9-,10+,11+/m0/s1
Formula Weight 198.1256
Chemical Name or Material Tacrolimus Methyl Acryl Aldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.