CAS RN 109466-74-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tacrolimus Methyl Acryl Aldehyde, TRC
CAS: 109466-74-2 Molecular Formula: C11 H18 O3 Molecular Weight (g/mol): 198.26 Synonym: 2-Propenal, 3-(4-hydroxy-3-methoxycyclohexyl)-2-methyl-, [1R-[1α(E),3α,4β]]- (9CI) IUPAC Name: (E)-3-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-2-methylprop-2-enal SMILES: CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)\C=O