missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ T-REx™ Tango™ GHSR-bla U2OS Cells

Product Code. 10595664
Click to view available options
Quantity:
8 x 106 Cells
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10595664

Brand: Thermo Scientific™ K1819

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The T-REx™ Tango™ GHSR-bla U2OS cells contain the human Growth Hormone Secretagogue Receptor (GHSR) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.

The T-REx™ Tango™ GHSR-bla U2OS cells contain the human Growth Hormone Secretagogue Receptor (GHSR) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.

The T-REx™ Tango™ GHSR-bla U2OS cells are functionally validated for Z' and EC50 concentrations of Ghrelin. In addition, T-REx™ Tango™ GHSR-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.

For Academic pricing please login or contact us at discoverysciences@invitrogen.com.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NM_198407
Assay Entry Cell-based beta-arrestin recruitment
Cell State Dividing Cells
Content And Storage LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.

Includes:
• T-REx™ Tango™ GHSR-bla U2OS cells
• T-REx™ Tango™ GHSR-bla U2OS Assay Protocol
• Certificate of Analysis

The T-REx™ Tango™ GHSR-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Cytoplasmic Tail Tail from AVPR2 GPCR
Description T-REx™ Tango™ GHSR-bla U2OS Cells, 8 x 106 Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
Detection Method Fluorescence
For Use With (Application) FRET
Gene Symbol GHSR
Human Protein Class G-Protein Coupled Receptor (GPCR)
Includes T-REx™ Tango™ GHSR-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: T-REx™ Tango™ GHSR-bla U2OS cells, T-REx™ Tango™ GHSR-bla U2OS assay protocol, certificate of analysis
Population Status Single Cell Clone Origin
Product Line GeneBLAzer, Tango
Quantity 8 x 106 Cells
Readout End Point
Sample Type U2OS
Target Entry GHSR
Type U2OS Cell
Cell Line U2OS
System Type Tango™
Druggable Target GPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.