missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet/TBX21 Antibody (4B10), PerCP/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
£251.00 - £492.00
Specifications
| Antigen | T-bet/TBX21 |
|---|---|
| Clone | 4B10 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000 |
| Applications | Flow Cytometry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18271924
|
Novus Biologicals
NBP1-42835-0.025MG |
0.025 mg |
£251.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18268403
|
Novus Biologicals
NBP1-42835-0.1MG |
0.1 mg |
£492.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Flow Cytometry.Specifications
| T-bet/TBX21 | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| PerCP-Cyanine5.5 | |
| RUO | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
| 30009 | |
| The immunogen for this antibody was TBet. | |
| Primary | |
| Store at 4C. Do not freeze. |
| 4B10 | |
| Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rhesus Macaque | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| TBX21 | |
| IgG1 κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title