missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet/TBX21 Antibody (4B10), PerCP/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-42835-0.025MG
This item is not returnable.
View return policy
Description
T-bet/TBX21 Monoclonal specifically detects T-bet/TBX21 in Human, Mouse, Rhesus Macaque samples. It is validated for Flow Cytometry.
Specifications
| T-bet/TBX21 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1:10-1:1000 | |
| T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 30009 | |
| Store at 4C. Do not freeze. |
| Flow Cytometry | |
| 4B10 | |
| PerCP-Cyanine5.5 | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
| TBX21 | |
| The immunogen for this antibody was TBet. | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rhesus Macaque | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction