missing translation for 'onlineSavingsMsg'
Learn More

(T-4)-Difluoro[2-(4-methoxyphenyl)-5-[2,2,2-trifluoro-1-[2-(4-methoxyphenyl)-5H-furo[3,2-b]pyrrol-5-ylidene-KappaN4]ethyl]-4H-furo[3,2-b]pyrrolato-KappaN4]boron, TRC

Product Code. 30386575
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386575 10 mg
30385124 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386575 Supplier Toronto Research Chemicals Supplier No. TRCD44623510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 956296-69-8
Molecular Formula C28H18BF5N2O4
Molecular Weight (g/mol) 552.26
Synonym 8-Desphenyl-8-trifluoromethyl Bodipy 8
SMILES COc1ccc(cc1)C2=CC3=[N+]4C(=C(c5cc6oc(cc6n5[B-]4(F)F)c7ccc(OC)cc7)C(F)(F)F)C=C3O2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C28H18BF5N2O4/c1-37-17-7-3-15(4-8-17)23-11-19-25(39-23)13-21-27(28(30,31)32)22-14-26-20(36(22)29(33,34)35(19)21)12-24(40-26)16-5-9-18(38-2)10-6-16/h3-14H,1-2H3
Formula Weight 552.128
Chemical Name or Material (T-4)-Difluoro[2-(4-methoxyphenyl)-5-[2,2,2-trifluoro-1-[2-(4-methoxyphenyl)-5H-furo[3,2-b]pyrrol-5-ylidene-kN4]ethyl]-4H-furo[3,2-b]pyrrolato-kN4]boron
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.