CAS RN 956296-69-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(T-4)-Difluoro[2-(4-methoxyphenyl)-5-[2,2,2-trifluoro-1-[2-(4-methoxyphenyl)-5H-furo[3,2-b]pyrrol-5-ylidene-KappaN4]ethyl]-4H-furo[3,2-b]pyrrolato-KappaN4]boron, TRC
CAS: 956296-69-8 Molecular Formula: C28H18BF5N2O4 Molecular Weight (g/mol): 552.26 Synonym: 8-Desphenyl-8-trifluoromethyl Bodipy 8 SMILES: COc1ccc(cc1)C2=CC3=[N+]4C(=C(c5cc6oc(cc6n5[B-]4(F)F)c7ccc(OC)cc7)C(F)(F)F)C=C3O2