missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT11 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | SYT11 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30228870
|
Novus Biologicals
NBP3-33397-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232703
|
Novus Biologicals
NBP3-33397-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SYT11 Monoclonal antibody specifically detects SYT11 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| SYT11 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neurotransmission | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 23208 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp781D015, KIAA0080synaptotagmin 12, MGC10881, MGC17226, synaptotagmin XISYT12, synaptotagmin-11, SytXI | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 37-200 of human SYT11 (SYT11). (NP_689493.3).,, Sequence:, WSCCHQQAEKKQKNPPYKFIHMLKGISIYPETLSNKKKIIKVRRDKDGPGREGGRRNLLVDAAEAGLLSRDKDPRGPSSGSCIDQLPIKMDYGEELRSPITSLTPGESKTTSPSSPEEDVMLGSLTFSVDYNFPKKALVVTIQEAHGLPVMDDQTQGSDPYIKM | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title