missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYT11 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33397-100ul
This item is not returnable.
View return policy
Description
SYT11 Monoclonal antibody specifically detects SYT11 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| SYT11 | |
| Monoclonal | |
| Western Blot 1:1000 - 1:5000, ELISA Recommended starting concentration is 1 μg/mL | |
| DKFZp781D015, KIAA0080synaptotagmin 12, MGC10881, MGC17226, synaptotagmin XISYT12, synaptotagmin-11, SytXI | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 37-200 of human SYT11 (SYT11). (NP_689493.3).,, Sequence:, WSCCHQQAEKKQKNPPYKFIHMLKGISIYPETLSNKKKIIKVRRDKDGPGREGGRRNLLVDAAEAGLLSRDKDPRGPSSGSCIDQLPIKMDYGEELRSPITSLTPGESKTTSPSSPEEDVMLGSLTFSVDYNFPKKALVVTIQEAHGLPVMDDQTQGSDPYIKM | |
| 100 μL | |
| Neuronal Cell Markers, Neurotransmission | |
| 23208 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction