missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntrophin gamma 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£385.00 - £587.00
Specifications
| Antigen | Syntrophin gamma 2 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389125
|
Novus Biologicals
NBP3-17441-25UL |
25 μg |
£385.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18373832
|
Bio-Techne
NBP3-17441-100UL |
100 μg |
£587.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Syntrophin gamma 2 Polyclonal antibody specifically detects Syntrophin gamma 2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Syntrophin gamma 2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| G2SYNgamma2-syntrophin, gamma-2-syntrophin, MGC133174, SYN5syntrophin 5, syntrophin, gamma 2, syntrophin-5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: ILRFYTAQDGTDWLRAVSANIRELTLQNMKMANKCCSPSDQVVHMGWVNEKLQGADSSQTFRPKFLALK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.2, 40% glycerol | |
| 54221 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title