missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNJ1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | SYNJ1 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16863165
|
Proteintech
24677-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16853165
|
Proteintech
24677-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4, 5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.Specifications
| SYNJ1 | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| O43426, Q8CHC4 | |
| 104015, 8867 | |
| SYNJ1 Fusion Protein Ag20364 | |
| Primary | |
| -20°C | |
| SYNJ1 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| INPP5G, KIAA0910, synaptojanin 1, SYNJ1 | |
| SYNJ1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title