missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNJ1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24677-1-AP-20UL
This item is not returnable.
View return policy
Description
Synaptojanin 1 is a phosphoinositide phosphatase that regulates levels of membrane phosphatidylinositol-4, 5-bisphosphate. As such, expression of this enzyme may affect synaptic transmission and membrane trafficking.Specifications
| SYNJ1 | |
| Polyclonal | |
| Unconjugated | |
| SYNJ1 | |
| INPP5G, KIAA0910, synaptojanin 1, SYNJ1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104015, 8867 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43426, Q8CHC4 | |
| SYNJ1 | |
| SYNJ1 Fusion Protein Ag20364 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction