missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader
GREENER_CHOICE

Product Code. 17228049 Shop All Agilent BioTek Products
Click to view available options
Model:
NEO2
NEO2ALPHABT
NEO2ALPHAT
NEO2M
NEO2MALPHA
NEO2MALPHAB
NEO2MALPHABT
NEO2MALPHAT
NEO2MB
NEO2MBT
NEO2MON
NEO2MT
NEO2S
NEO2SALPHA
NEO2SM
NEO2SMALPHA
NEO2SMALPHAB
NEO2SMB
NEO2T
Unit Size:
1 unit
Each
This item is not returnable. View return policy

Product Code. 17228049

Brand: Agilent BioTek N2SMBSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.

BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.

Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range 230 to 999 nm, 1 nm increments
Accuracy Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD
Certifications/Compliance CE and TUV marked. RoHS compliant.
CO2 Concentration Range 0 to 20%
Depth (Metric) 52.5 cm
Depth (English) 20.7 in.
Incubator Temperature ±0.2°C at 37°C
Shaking Linear, Orbital, Double Orbital
Dynamic Range 0 to 4 OD
Dispense Volume 5 to 1000 μL, 1 μL increments
For Use With (Application) HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP
Height (English) 16.1 in.
Height (Metric) 41 cm
Linearity ≤1% from 0 to 3 OD
Includes Monochromater: ABS, FI and LUM; Filter: Single filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF
Model NEO2SMB
Product Type Multi-Mode Microplate Reader
Power Consumption 250 W max.
Resolution 0.0001 OD
Weight (English) 78 lb.
Weight (Metric) 35 kg
Width (English) 15.4 in.
Width (Metric) 39 cm
Detector Type Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option)
Software Type Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Light Source Xenon Flash
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.