Learn More
Agilent BioTek Synergy HTX Multi-Mode Microplate Reader, IVD
Shop All Agilent BioTek Products

Description
The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows
With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.
Specifications
Specifications
| Absorbance Range | 200 to 999 nm, 1 nm increments |
| Accuracy | Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2% |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Depth (Metric) | 38 cm |
| Compatibility | BioStack and Third Party Automation Compatible |
| Depth (English) | 15 in. |
| Incubator Temperature | 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C) |
| Measurement Speed | 31 sec. (96 wells), 80 sec. (384 wells) |
| Shaking | Linear, Orbital |
| Dynamic Range | Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
