missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek Synergy HTX Multi-Mode Microplate Reader
GREENER_CHOICE

Product Code. 17288049 Shop All Agilent BioTek Products
Click to view available options
Model:
S1A
S1L
S1LA
S1LF
S1LFA
S1LFTA
Includes:
Absorbance, Incubation, Shaking
Absorbance, Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking
Absorbance, Luminescence, Fluorescence (Top/Bottom), Time-resolved Fluorescence (TRF - Secondary Mode), Alpha, Incubation, Shaking
Absorbance, Luminescence, Incubation, Shaking
Luminescence, Fluorescence (Top/Bottom), Alpha, Incubation, Shaking
Luminescence, Incubation, Shaking
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17288049

Brand: Agilent BioTek S1LSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. For Research Use Only. Not for use in diagnostic procedures.

The BioTek Synergy HTX is a compact, economical multi-mode reader for 6- to 384-well microplates and Take3 microvolume plates. The unique dual optics design provides superior performance for UV-Vis absorbance, fluorescence, luminescence, and AlphaScreen/AlphaLISA workflows

With incubation and shaking, plus a dual reagent injector module, the system will meet all your assay requirements now and in the future. Synergy HTX is controlled by the easy-to-use, yet powerful, Gen5 software for data collection, analysis, exporting, and reporting.

Order Info

Optional Accessories: Dual Reagent Dispenser Module, Gen5 Secure (for 21 CFR Part 11 Compliance), Fluorescence Test Plate, Absorbance Test Plate, Luminescence Test Plate, Patented BioCell 1 cm quartz vessel, Product Qualification Package, Take3/Take3 Trio.

Certifications

CE and TUV marked

Compliance

CE and TUV marked

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Range 200 to 999 nm, 1 nm increments
Accuracy Absorbance: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD; Dispense: ±1 μL or 2%
Certifications/Compliance CE and TUV marked. RoHS compliant.
Depth (Metric) 38 cm
Compatibility BioStack and Third Party Automation Compatible
Depth (English) 15 in.
Incubator Temperature 4-Zone Incubation to 50°C with Condensation Control (±0.2°C at 37°C)
Measurement Speed 31 sec. (96 wells), 80 sec. (384 wells)
Shaking Linear, Orbital
Dynamic Range Absorbance: 0 to 4 OD; Fluorescence Intensity/Luminesence: >6 decades
Height (English) 10 in.
Frequency 50/60 Hz
Height (Metric) 25.4 cm
Includes Luminescence, Incubation, Shaking
Model S1L
No. of Dispensers 2 Syringe Pumps
Product Type Multi-Mode Microplate Reader
USB Connections 2
Voltage 100/240 V
Weight (English) 40 lb.
Weight (Metric) 18 kg
Width (English) 16 in.
Width (Metric) 40.6 cm
Plate Type 6- to 384-well Plates
Detector Type Photodiode, PMT
Software Type Gen5 Microplate Reader and Imager Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Light Source Xenon Flash, Tungsten Halogen (Alpha Detection/Fluorescence Intensity)
Reading Options End Point, Kinetic, Spectral Scanning, Well Area Scanning
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.