missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SYNE3 Recombinant Rabbit Monoclonal Antibody (23GB4670), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553784
This item is not returnable.
View return policy
Description
As a component of the LINC (LInker of Nucleoskeleton and Cytoskeleton) complex involved in the connection between the nuclear lamina and the cytoskeleton. The nucleocytoplasmic interactions established by the LINC complex play an important role in the transmission of mechanical forces across the nuclear envelope and in nuclear movement and positioning. Probable anchoring protein which tethers the nucleus to the cytoskeleton by binding PLEC which can associate with the intermediate filament system. Plays a role in the regulation of aortic epithelial cell morphology, and is required for flow-induced centrosome polarization and directional migration in aortic endothelial cells. [UniProt].
Specifications
| SYNE3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SYNE3 | |
| C14orf139; C14orf49; KASH domain-containing protein 3; KASH3; LINC00341; Nesp3; nesprin-3; NET53; Nuclear envelope spectrin repeat protein 3; RGD1304563; spectrin repeat containing nuclear envelope family member 3; spectrin repeat containing, nuclear envelope family member 3; SYNE3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 161176 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4670 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ZMZ3 | |
| SYNE3 | |
| A synthesized peptide derived from human Nesprin3 (750-935AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction