missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYNE3 Recombinant Rabbit Monoclonal Antibody (23GB4670), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£179.00 - £485.00

Specifications

Antigen SYNE3
Clone 23GB4670
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089590
GREENER_CHOICE
View Documents
Invitrogen™
MA553784
100 μL
£485.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30089606
View Documents
Invitrogen™
MA553785
20 μL
£179.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SYNE3 Recombinant Monoclonal antibody specifically detects SYNE3 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications

Specifications

SYNE3
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human
Q6ZMZ3
161176
IgG
Affinity chromatography
Antibody
23GB4670
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
C14orf139; C14orf49; KASH domain-containing protein 3; KASH3; LINC00341; Nesp3; nesprin-3; NET53; Nuclear envelope spectrin repeat protein 3; RGD1304563; spectrin repeat containing nuclear envelope family member 3; spectrin repeat containing, nuclear envelope family member 3; SYNE3
A synthesized peptide derived from human Nesprin3 (750-935AA).
Primary
-20°C
SYNE3
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.