missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNE1 Antibody (6A), Novus Biologicals™
Mouse Monoclonal Antibody
£155.00
Specifications
| Antigen | SYNE1 |
|---|---|
| Clone | 6A |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18743014
|
Novus Biologicals
NBP2-22477-0.025mg |
0.025 mg | |||||||
Description
SYNE1 Monoclonal specifically detects SYNE1 in Rat samples. It is validated for Western Blot.Specifications
| SYNE1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Developmental Biology | |
| ARCA1, C6orf98, CPG2, dJ45H2.2, EC 4.2.1.49, EC 4.3.1.3, EDMD4, Enaptin, KIAA0796chromosome 6 open reading frame 98, KIAA1262FLJ41140, KIAA17568B, Myne-1, MYNE1DKFZp781J13156, Myocyte nuclear envelope protein 1, nesprin 1, nesprin-1, Nuclear envelope spectrin repeat protein 1, SCAR8FLJ30878, spectrin repeat containing, nuclear envelope 1, Synaptic nuclear envelope protein 1, synaptic nuclei expressed gene 1, Syne-1, SYNE-1B | |
| SYNE1 | |
| IgG1 | |
| Protein G purified | |
| This antibody reacts with Rat. |
| 6A | |
| Western Blot 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Rat | |
| 23345 | |
| Recombinant protein containing residues RKLESTLTGLEQSRERQERR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title