missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNE1 Antibody (6A), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22477-0.025mg
This item is not returnable.
View return policy
Description
SYNE1 Monoclonal specifically detects SYNE1 in Rat samples. It is validated for Western Blot.
Specifications
| SYNE1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| SYNE1 | |
| Recombinant protein containing residues RKLESTLTGLEQSRERQERR | |
| 0.025 mg | |
| Developmental Biology | |
| 23345 | |
| Rat | |
| Purified |
| Western Blot | |
| 6A | |
| Unconjugated | |
| ARCA1, C6orf98, CPG2, dJ45H2.2, EC 4.2.1.49, EC 4.3.1.3, EDMD4, Enaptin, KIAA0796chromosome 6 open reading frame 98, KIAA1262FLJ41140, KIAA17568B, Myne-1, MYNE1DKFZp781J13156, Myocyte nuclear envelope protein 1, nesprin 1, nesprin-1, Nuclear envelope spectrin repeat protein 1, SCAR8FLJ30878, spectrin repeat containing, nuclear envelope 1, Synaptic nuclear envelope protein 1, synaptic nuclei expressed gene 1, Syne-1, SYNE-1B | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody reacts with Rat. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction