missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptojanin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£280.00 - £459.00
Specifications
| Antigen | Synaptojanin 1 |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500, KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18410382
|
Novus Biologicals
NBP1-87842-25ul |
25 μL |
£280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18758553
|
Novus Biologicals
NBP1-87842 |
0.1 mL |
£459.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synaptojanin 1 Polyclonal specifically detects Synaptojanin 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Synaptojanin 1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.1.3, EC 3.1.3.36, KIAA0910, polyphosphoinositide phosphatase, 10synaptojanin-1, Synaptic inositol-14,5-trisphosphate 5-phosphatase 1, synaptojanin 1 | |
| SYNJ1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human Synaptojanin 1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500, KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 8867 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ASFGEVILIRFVEDKMWVTFLEGSSALNVLSLNGKELLNRTITIALKSPDWIKNLEEEMSLEKISIALPSSTSSTLLGEDAEVAADFDMEGDVDDYSAEVEELLPQHLQPSSSSGLGTSPSSSPRTSPCQS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title