Learn More
Synaptojanin 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-87842
This item is not returnable.
View return policy
Description
Synaptojanin 1 Polyclonal specifically detects Synaptojanin 1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500, KnockDown Validated | |
| Affinity Purified | |
| RUO |
| Polyclonal | |
| SYNJ1 | |
| 0.1 mL | |
| IgG |