missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synapsin II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £395.00
Specifications
| Antigen | Synapsin II |
|---|---|
| Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:100 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18280095
|
Novus Biologicals
NB120-13258-0.025MG |
0.025 mg |
£155.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284980
|
Novus Biologicals
NB120-13258 |
0.1 mg |
£395.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synapsin II Polyclonal specifically detects Synapsin II in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Synapsin II | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 6854 | |
| Synthetic peptide corresponding to a portion of rat Synapsin II. The sequence is completely conserved in mouse. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat, Mouse | |
| synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb | |
| SYN2 | |
| IgG | |
| Protein A purified | |
| This detects ∼74 and ∼54 kDa proteins, corresponding to the apparent molecular masses of synapsin IIa and IIb on SDS-PAGE immunoblots, in samples from mouse and rat origins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title