missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synapsin II Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-13258-0.025MG
This item is not returnable.
View return policy
Description
Synapsin II Polyclonal specifically detects Synapsin II in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Synapsin II | |
| Polyclonal | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:100 | |
| synapsin IISYNII, synapsin-2, SYNIIa, SYNIIb | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This detects ∼74 and ∼54 kDa proteins, corresponding to the apparent molecular masses of synapsin IIa and IIb on SDS-PAGE immunoblots, in samples from mouse and rat origins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| SYN2 | |
| Synthetic peptide corresponding to a portion of rat Synapsin II. The sequence is completely conserved in mouse. | |
| 0.025 mg | |
| Neuroscience | |
| 6854 | |
| Rat, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction