missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYDE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57607
This item is not returnable.
View return policy
Description
SYDE1 Polyclonal specifically detects SYDE1 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| SYDE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 2.4.1.25, EC 2.6.1.9, FLJ13511, Protein syd-1 homolog 1, rho GTPase-activating protein SYDE1, synapse defective 1, Rho GTPase, homolog 1 (C. elegans), Synapse defective protein 1 homolog 1,7h3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 85360 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Knockout Validated | |
| Q6ZW31 | |
| SYDE1 | |
| Synthetic peptides corresponding to SYDE1(synapse defective 1, Rho GTPase, homolog 1 (C. elegans)) The peptide sequence was selected from the C terminal of SYDE1. Peptide sequence PYLRPKRQPPLHLPLADPEVVTRPRGRGGPESPPSNRYAGDWSVCGRDFL. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Pig: 92%; Canine: 84%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction