missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For use with Eppendorf Centrifuges 5810/5810R
- Centrifuge 5810/5810 R is a workhorse for medium to high-throughput laboratories. It combines extraordinary versatility and capacity for both tubes and plates with an extraordinary compact footprint.
Specifications
Specifications
| Type | Swinging Bucket Rotor |
| Capacity | 0.2 to 500 mL |
| Max. RCF | 3,220 x g |
| Max. Speed | 4,000 rpm |
| For Use With (Equipment) | 5810/5810R Centrifuge |
| Includes | Aerosol-tight caps for safe centrifugation of hazardous samples optionally available |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?