missing translation for 'onlineSavingsMsg'
Learn More

Bolle Safety SWIFT Safety Eyewear with Platinum Lite Anti-fog and Anti-scratch Coating

Product Code. 31810815 Shop All Bollé Safety Products
Change view
Click to view available options
Frame Color:
Blue
Lens Tint:
Copper
Smoke
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Color Lens Tint Unit Size
31810815 Blue Smoke Each
31810790 Blue Copper Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31810815 Supplier Bolle Safety Supplier No. SWIFTN20E
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Delivers reliable everyday protection with clear vision and lasting comfort.

  • Reliable everyday protection: Designed to meet essential safety standards for consistent eye protection across a wide range of work environments.
  • PLATINUM Lite coating technology: Anti-fog and anti-scratch coating ensures clear, reliable vision in all working conditions.
  • Crystal-clear visibility: Maintains high optical clarity to support precision tasks and reduce the risk of errors.
  • All-day comfort: Well-balanced frame minimizes pressure points, enhancing comfort during extended wear.
  • Textured temples for better retention: Improves grip and stability, even during active movements.
  • Optimized lateral protection: Temple design enhances side coverage, increasing overall eye safety.
  • Durable construction: Engineered for regular use, helping extend product lifespan and reduce replacement needs.
  • Eco-designed product: Made from 31% recycled polycarbonate, supporting sustainability initiatives.
  • Supports ESG objectives: Helps companies reduce environmental impact while maintaining high safety standards.
  • Encourages wearer compliance: Comfortable, lightweight, and practical design promotes consistent use.
  • Versatile applications: Suitable for manufacturing, logistics, maintenance, construction, and general industry.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN ISO 16321-1
Lens Type C-Rated (Impact), GL3 Rated
Lens Coating Platinum Lite, Anti-fog, Anti-scratch
Lens Tint Smoke
Product Type Safety Glasses
Frame Color Blue
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV380
Frame Type Frameless
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.