missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 Small T antigen Antibody (pab 280) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61248
This item is not returnable.
View return policy
Description
SV40 Small T antigen Monoclonal specifically detects SV40 Small T antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| SV40 Small T antigen | |
| Monoclonal | |
| 0.9 mg/ml | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Mouse | |
| Protein A purified | |
| Cancer, Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| pab 280 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| A purified Pab 419-SV40 Small T antigen | |
| 0.1 mL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction