missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 Small T antigen Antibody (pab 280) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£159.00 - £302.00
Specifications
| Antigen | SV40 Small T antigen |
|---|---|
| Clone | pab 280 |
| Concentration | 0.9 mg/ml |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18296654
|
Novus Biologicals
NBP2-61248 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635358
|
Novus Biologicals
NBP2-61248-0.025ml |
0.025 mL |
£159.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SV40 Small T antigen Monoclonal specifically detects SV40 Small T antigen in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| SV40 Small T antigen | |
| 0.9 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Virus | |
| A purified Pab 419-SV40 Small T antigen | |
| Primary |
| pab 280 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| Cancer, Virology Bacteria and Parasites | |
| PBS with 0.02% Sodium Azide | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title