missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUV420h1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £375.00
Specifications
| Antigen | SUV420h1 |
|---|---|
| Dilution | Western Blot 1:200-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18604932
|
Novus Biologicals
NBP2-94791-0.02ml |
0.02 mL |
£159.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661212
|
Novus Biologicals
NBP2-94791-0.1ml |
0.1 mL |
£375.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUV420h1 Polyclonal antibody specifically detects SUV420h1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| SUV420h1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Epigenetics, Transcription Factors and Regulators | |
| PBS (pH 7.3), 50% glycerol | |
| 51111 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C630029K18Rik, CGI85, CGI-85, EC 2.1.1.43, histone-lysine N-methyltransferase SUV420H1, KMT5B, KMT5B Lysine N-methyltransferase 5B, MGC118906, MGC118909, MGC21161, MGC703, Su(var)4-20 homolog 1, Suppressor of variegation 4-20 homolog 1, suppressor of variegation 4-20 homolog 1 (Drosophila), suv4-20h1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SUV420H1 (NP_060105.3). MNTSAFPSRSSRHFSKSDSFSHNNPVRFRPIKGRQEELKEVIERFKKDEHLEKAFKCLTSGEWARHYFLNKNKMQEKLFKEHVFIYLRMFATDSGFEILPC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title