missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUR1 Antibody (S289-16), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
£176.00 - £477.00
Specifications
| Antigen | SUR1 |
|---|---|
| Clone | S289-16 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18081906
|
Novus Biologicals
NBP2-59320 |
100 μg |
£477.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698608
|
Novus Biologicals
NBP2-59320-25ug |
25 μg |
£176.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUR1 Monoclonal specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| SUR1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| Unconjugated | |
| Mouse | |
| ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain | |
| ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8 | |
| ABCC8 | |
| IgG1 | |
| Protein G purified |
| S289-16 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Hamster | |
| 6833 | |
| Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1 | |
| Primary | |
| Detects 160kDa. Does not cross-react with SUR2B. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title