missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUR1 Antibody (S289-16), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-59320
This item is not returnable.
View return policy
Description
SUR1 Monoclonal specifically detects SUR1 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SUR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.09% Sodium Azide | |
| ABCC8 | |
| Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1 | |
| 100 μg | |
| ABC Transporters, Lipid and Metabolism, Nutrient Sensing in the Brain | |
| 6833 | |
| Human, Mouse, Rat, Hamster | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S289-16 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| ABC36, ATP-binding cassette, sub-family C (CFTR/MRP), member 8, HHF1Sulfonylurea receptor 1, HI, PHHIHRINS, sulfonylurea receptor (hyperinsulinemia), SUR1MRP8, SURATP-binding cassette transporter sub-family C member 8, TNDM2ATP-binding cassette sub-family C member 8 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects 160kDa. Does not cross-react with SUR2B. | |
| Store at -20C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction